missing translation for 'onlineSavingsMsg'
Learn More

N-[2-(Methylamino)ethyl]-5-isoquinolinesulfonamide Dihydrochloride, TRC

Product Code. 30469887
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30469887 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30469887 Supplier Toronto Research Chemicals Supplier No. TRCM2862005MG
 Restricted Item
Estimated Shipment: 24-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N-[2-(Methylamino)ethyl]-5-isoquinolinesulfonamide','Dihydroc
CAS 113276-94-1
Molecular Formula C12 H15 N3 O2 S . 2 Cl H
Quantity 5 mg
InChI Formula InChI=1S/C12H15N3O2S.2ClH/c1-13-7-8-15-18(16,17)12-4-2-3-10-9-14-6-5-11(10)12;;/h2-6,9,13,15H,7-8H2,1H3;2*1H
SMILES Cl.Cl.CNCCNS(=O)(=O)c1cccc2cnccc12
IUPAC Name N-[2-(methylamino)ethyl]isoquinoline-5-sulfonamide;dihydrochloride
Molecular Weight (g/mol) 338.25
Formula Weight 337.0419
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.