Learn More
N-2 Trityl Olmesartan Medoxomil, TRC
Product Code. 30449036
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30449036 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30449036 Supplier Toronto Research Chemicals Supplier No. TRCT889420100MG
 Restricted Item
Estimated Shipment: 10-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material N-2 Trityl Olmesartan Medoxomil
CAS 1020157-01-0
Molecular Formula C48 H44 N6 O6
Quantity 100 mg
Synonym 1H-Imidazole-5-carboxylic acid','4-(1-hydroxy-1-methylethyl)-2-propyl-1-[[2'-[2-(triphenylmethyl)-2H-tetrazol-5-yl][1,1'-biphenyl]-4-yl]methyl]-','(5-methyl-2-oxo-1,3-dioxol-4-yl)methyl ester, (5-Methyl-2-oxo-1,3-dioxol-4-yl)methyl 4-(1-Hydroxy-1-methylethyl)-2-propyl-1-[[2'-[(2-triphenylmethyl)-2H-tetrazol-5-yl]biphenyl-4-yl]methyl]-1H-imidazole-5-carboxylate, N-2 Trityl Olmesartan Medoxomil
InChI Formula InChI=1S/C48H44N6O6/c1-5-17-41-49-43(47(3,4)57)42(45(55)58-31-40-32(2)59-46(56)60-40)53(41)30-33-26-28-34(29-27-33)38-24-15-16-25-39(38)44-50-52-54(51-44)48(35-18-9-6-10-19-35,36-20-11-7-12-21-36)37-22-13-8-14-23-37/h6-16,18-29,57H,5,17,30-31H2,1-4H3
SMILES CCCc1nc(c(C(=O)OCC2=C(C)OC(=O)O2)n1Cc3ccc(cc3)c4ccccc4c5nnn(n5)C(c6ccccc6)(c7ccccc7)c8ccccc8)C(C)(C)O
IUPAC Name (5-methyl-2-oxo-1,3-dioxol-4-yl)methyl 5-(2-hydroxypropan-2-yl)-2-propyl-3-[[4-[2-(2-trityltetrazol-5-yl)phenyl]phenyl]methyl]imidazole-4-carboxylate
Molecular Weight (g/mol) 800.9
Formula Weight 800.3322