missing translation for 'onlineSavingsMsg'
Learn More

N-[3-(3-Cyanopyrazolo[1,5-a]pyrimidin-7-yl)phenyl]acetamide, TRC

Product Code. 17942625
Change view
Click to view available options
Quantity:
10 mg
100 mg
Unit Size:
100mg
10mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17942625 10 mg 10mg
17925005 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17942625 Supplier Toronto Research Chemicals Supplier No. TRCC98767010MG
 Restricted Item
Estimated Shipment: 09-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 115931-01-6
Molecular Formula C15 H11 N5 O
Molecular Weight (g/mol) 277.28
Synonym CL 284859
IUPAC Name N-[3-(3-cyanopyrazolo[1,5-a]pyrimidin-7-yl)phenyl]acetamide
SMILES CC(=O)Nc1cccc(c1)c2ccnc3c(cnn23)C#N

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C15H11N5O/c1-10(21)19-13-4-2-3-11(7-13)14-5-6-17-15-12(8-16)9-18-20(14)15/h2-7,9H,1H3,(H,19,21)
Formula Weight 277.0964
Chemical Name or Material N-[3-(3-Cyanopyrazolo[1,5-a]pyrimidin-7-yl)phenyl]acetamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.