missing translation for 'onlineSavingsMsg'
Learn More

N-(3,5-Dimethyl-2(3H)-thiazolylidene)-4-hydroxy-2-methyl-2H-1,2-benzothiazine-3-carboxamide 1,1-Dioxide, TRC

Product Code. 30137584
Change view
Click to view available options
Quantity:
100 mg
250 mg
Unit Size:
100mg
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30137584 250 mg 250mg
30012389 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30137584 Supplier Toronto Research Chemicals Supplier No. TRCD479479250MG
 Restricted Item
Estimated Shipment: 28-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1145656-36-5
Molecular Formula C15 H15 N3 O4 S2
Molecular Weight (g/mol) 365.43
Synonym 2H-1,2-Benzothiazine-3-carboxamide, N-(3,5-dimethyl-2(3H)-thiazolylidene)-4-hydroxy-2-methyl-, 1,1-dioxide, N-[3,5-Dimethylthiazol-2(3H)-ylidene]-4-hydroxy-2-methyl-2H-1,2-benzothiazine-3-carboxamide 1,1-dioxide, (EZ)-N'-Methylmeloxicam, (EZ)-N-(3,5-dimethylthiazol-2(3H)-ylidene)-4-hydroxy-2-methyl-2H-benzo[e][1,2]thiazine-3-carboxamide 1,1-dioxide
IUPAC Name N-(3,5-dimethyl-1,3-thiazol-2-ylidene)-4-hydroxy-2-methyl-1,1-dioxo-1λ^{6},2-benzothiazine-3-carboxamide
SMILES CN1C=C(C)SC1=NC(=O)C2=C(O)c3ccccc3S(=O)(=O)N2C

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C15H15N3O4S2/c1-9-8-17(2)15(23-9)16-14(20)12-13(19)10-6-4-5-7-11(10)24(21,22)18(12)3/h4-8,19H,1-3H3
Formula Weight 365.0504
Chemical Name or Material (Z)-N'-Methylmeloxicam
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.