missing translation for 'onlineSavingsMsg'
Learn More

(+/-)-N-3-Benzylnirvanol, TRC

Product Code. 30133027
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30133027 50 mg 50mg
30004964 10 mg 10mg
30007094 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30133027 Supplier Toronto Research Chemicals Supplier No. TRCB28577050MG
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 93879-40-4
Molecular Formula C18 H18 N2 O2
Molecular Weight (g/mol) 294.35
IUPAC Name 3-benzyl-5-ethyl-5-phenylimidazolidine-2,4-dione
SMILES CCC1(NC(=O)N(Cc2ccccc2)C1=O)c3ccccc3

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C18H18N2O2/c1-2-18(15-11-7-4-8-12-15)16(21)20(17(22)19-18)13-14-9-5-3-6-10-14/h3-12H,2,13H2,1H3,(H,19,22)
Formula Weight 294.1368
Percent Purity >95%
Chemical Name or Material (+/-)-N-3-Benzylnirvanol
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.