missing translation for 'onlineSavingsMsg'
Learn More

N-(3-Chloro-2-methylphenyl)-2-hydroxynicotinamide, TRC

Product Code. 17918504
Change view
Click to view available options
Quantity:
250 mg
2.5 g
Unit Size:
2.50g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17918504 250 mg 250mg
17989434 2.5 g 2.50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17918504 Supplier Toronto Research Chemicals Supplier No. TRCC369440250MG
 Restricted Item
Estimated Shipment: 06-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 72646-00-5
Molecular Formula C13 H11 Cl N2 O2
Molecular Weight (g/mol) 262.69
Synonym N-(3-Chloro-2-methylphenyl)-1,2-dihydro-2-oxo-3-pyridinecarboxamide, N-(3-Chloro-2-methylphenyl)-2-oxo-1,2-dihydropyridine-3-carboxamide
IUPAC Name N-(3-chloro-2-methylphenyl)-2-hydroxypyridine-3-carboxamide
SMILES Cc1c(Cl)cccc1NC(=O)c2cccnc2O

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C13H11ClN2O2/c1-8-10(14)5-2-6-11(8)16-13(18)9-4-3-7-15-12(9)17/h2-7H,1H3,(H,15,17)(H,16,18)
Formula Weight 262.0509
Chemical Name or Material N-(3-Chloro-2-methylphenyl)-2-hydroxynicotinamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.