Learn More
N-(3-Chloro-2-methylphenyl)-2-hydroxynicotinamide, TRC
Product Code. 17989434
Change view
Click to view available options
Quantity:
250 mg
2.5 g
Unit Size:
2.50g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17989434 2.5 g 2.50g
17918504 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 17989434 Supplier Toronto Research Chemicals Supplier No. TRCC3694402.5G
 Restricted Item
Estimated Shipment: 28-09-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 72646-00-5
Molecular Formula C13 H11 Cl N2 O2
Molecular Weight (g/mol) 262.69
Synonym N-(3-Chloro-2-methylphenyl)-1,2-dihydro-2-oxo-3-pyridinecarboxamide, N-(3-Chloro-2-methylphenyl)-2-oxo-1,2-dihydropyridine-3-carboxamide
IUPAC Name N-(3-chloro-2-methylphenyl)-2-hydroxypyridine-3-carboxamide
SMILES Cc1c(Cl)cccc1NC(=O)c2cccnc2O
Specifications
Quantity 2.5 g
InChI Formula InChI=1S/C13H11ClN2O2/c1-8-10(14)5-2-6-11(8)16-13(18)9-4-3-7-15-12(9)17/h2-7H,1H3,(H,15,17)(H,16,18)
Formula Weight 262.0509
Chemical Name or Material N-(3-Chloro-2-methylphenyl)-2-hydroxynicotinamide
Product Title
Seleccione un problema
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.