missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-(3-Phenylpropionyl)glycine-2,2-d2, CDN
Click to view available options
Quantity:
0.01 g
0.05 g
Unit Size:
0.01g
0.05g
Specifications
Specifications
| Chemical Name or Material | N-(3-Phenylpropionyl)glycine-2,2-D2 |
| CAS | 1219795-43-3 |
| Molecular Formula | C11 D2 H11 N O3 |
| Quantity | 0.05 g |
| Synonym | N-(1-Oxo-3-phenylpropyl)glycine-2,2-d2, (3-Phenylpropionyl)glycine-d2, 2-(3-Phenylpropanamido)acetic-2,2-d2 acid, N-(3-Phenylpropanoyl)glycine-2,2,-d2, N-(3-Phenylpropionyl)glycine-d2, N-(3-Phenylpropionyl)glycine-2,2-d2, N-(3-Phenylpropionyl)glycine-2,2-D2, [(3-Phenylpropionyl)amino]acetic-2,2-d2 acid |
| InChI Formula | InChI=1 S/C11H13NO3/c13-10(12-8-11(14)15)7-6-9-4-2-1-3-5-9/h1-5 H,6-8H2,(H,12,13)(H,14,15)/i8D2 |
| SMILES | [2 H]C([2 H])(NC(=O)CCc1ccccc1)C(=O)O |
| IUPAC Name | 2,2-dideuterio-2-(3-phenylpropanoylamino)acetic acid |
| Molecular Weight (g/mol) | 209.238 |
| Formula Weight | 209.102 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction