missing translation for 'onlineSavingsMsg'
Learn More

N-(4-Acetyl-3-methylphenyl)acetamide, 95%, BLD Pharm™

Product Code. 30963554
Encompass_Preferred
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
25 g
Unit Size:
1g
250mg
25g
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30963554 250 mg 250mg
30958488 5 g 5g
30950326 25 g 25g
30961860 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 30963554 Supplier BLD Pharm Supplier No. BD13196250mg
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 34956-31-5
Molecular Formula C11H13NO2
MDL Number MFCD02258876
Quantity 250 mg
SMILES CC(NC1=CC=C(C(C)=O)C(C)=C1)=O
Molecular Weight (g/mol) 191.23
Percent Purity 95%
Chemical Name or Material N-(4-Acetyl-3-methylphenyl)acetamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.