missing translation for 'onlineSavingsMsg'
Learn More

N-(4-Aminobenzenesulfonyl) Sulfamethoxazole, TRC

Product Code. 30134648
Change view
Click to view available options
Quantity:
100 mg
1 g
Unit Size:
100mg
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30134648 1 g 1g
30006096 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30134648 Supplier Toronto Research Chemicals Supplier No. TRCA5914801G
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 135529-16-7
Molecular Formula C16 H16 N4 O5 S2
Molecular Weight (g/mol) 408.45
Synonym 4-[[(4-Aminophenyl)sulphonyl]amino]-N-(5-methylisoxazol-3-yl)benzenesulphonamide, Sulfamethoxazole Imp. B (EP)
IUPAC Name 4-amino-N-[4-[(5-methyl-1,2-oxazol-3-yl)sulfamoyl]phenyl]benzenesulfonamide
SMILES Cc1onc(NS(=O)(=O)c2ccc(NS(=O)(=O)c3ccc(N)cc3)cc2)c1

Specifications

Quantity 1 g
InChI Formula InChI=1S/C16H16N4O5S2/c1-11-10-16(18-25-11)20-27(23,24)15-8-4-13(5-9-15)19-26(21,22)14-6-2-12(17)3-7-14/h2-10,19H,17H2,1H3,(H,18,20)
Formula Weight 408.0562
Percent Purity >95%
Chemical Name or Material 4-[((4-Aminophenyl)sulfonyl)amino]-N-(5-methylisoxazol-3-yl)benzenesulfonamide
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.