missing translation for 'onlineSavingsMsg'
Learn More

N-(4-Aminophenyl)-N-methyl-2-(4-methylpiperazin-1-yl)acetamide, TRC

Product Code. 17939464
Change view
Click to view available options
Quantity:
100 mg
250 mg
Unit Size:
100mg
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17939464 250 mg 250mg
17939204 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17939464 Supplier Toronto Research Chemicals Supplier No. TRCA628740250MG
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 262368-30-9
Molecular Formula C14H22N4O
Molecular Weight (g/mol) 262.35
Synonym N-(4-Aminophenyl)-N,4-dimethyl-1-piperazineacetamide, N-[[(4-Methylpiperazin-1-yl)methyl]carbonyl]-N-methyl-p-phenylenediamine
IUPAC Name N-(4-aminophenyl)-N-methyl-2-(4-methylpiperazin-1-yl)acetamide
SMILES CN1CCN(CC1)CC(=O)N(C)C2=CC=C(C=C2)N

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C14H22N4O/c1-16-7-9-18(10-8-16)11-14(19)17(2)13-5-3-12(15)4-6-13/h3-6H,7-11,15H2,1-2H3
Formula Weight 262.1794
Chemical Name or Material N-(4-Aminophenyl)-N-methyl-2-(4-methylpiperazin-1-yl)acetamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.