missing translation for 'onlineSavingsMsg'
Learn More

N-(4-Chlorophenyl)-1,3,5-triazine-2,4-diamine, TRC

Product Code. 30134693
Change view
Click to view available options
Quantity:
250 mg
500 mg
1 g
Unit Size:
1g
250mg
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30134693 500 mg 500mg
30138488 1 g 1g
30011831 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30134693 Supplier Toronto Research Chemicals Supplier No. TRCC424325500MG
 Restricted Item
Estimated Shipment: 04-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 500-42-5
Molecular Formula C9 H8 Cl N5
Molecular Weight (g/mol) 221.65
Synonym 1,3,5-Triazine-2,4-diamine, N2-(4-chlorophenyl)-, 1,3,5-Triazine-2,4-diamine, N-(4-chlorophenyl)- (9CI), s-Triazine, 2-amino-4-(p-chloroanilino)- (6CI,7CI,8CI), N2-(4-Chlorophenyl)-1,3,5-triazine-2,4-diamine, 2-(p-Chloroanilino)-4-amino-s-triazine, 2-Amino-4-(p-chloroanilino)-1,3,5-triazine, 2-Amino-4-p-chloroanilino-s-triazine, ASA 226, Chlorazanil, Chlorazinil, Chloroazinal, Diurazine, Diurazyna, N-(p-Chlorophenyl)-2,4-diamino-s-triazine, Neo-Urofort, Neurofort, Orpizin, Riker 545, SKF 3195, Triazurol, neo-Urofort
IUPAC Name 2-N-(4-chlorophenyl)-1,3,5-triazine-2,4-diamine
SMILES Nc1ncnc(Nc2ccc(Cl)cc2)n1

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C9H8ClN5/c10-6-1-3-7(4-2-6)14-9-13-5-12-8(11)15-9/h1-5H,(H3,11,12,13,14,15)
Formula Weight 221.0468
Chemical Name or Material N-(4-Chlorophenyl)-1,3,5-triazine-2,4-diamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.