missing translation for 'onlineSavingsMsg'
Learn More

N-(5-Amino-2-methylphenyl)-4-(3-pyridyl)-2-pyrimidineamine, TRC

Product Code. 17992175
Change view
Click to view available options
Quantity:
1 g
2.5 g
5 g
Unit Size:
1g
2.5g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17992175 5 g 5g
17981955 2.5 g 2.5g
17963355 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17992175 Supplier Toronto Research Chemicals Supplier No. TRCA6182205G
 Restricted Item
Estimated Shipment: 28-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 152460-10-1
Molecular Formula C16 H15 N5
Molecular Weight (g/mol) 277.32
Synonym N-(5-Amino-2-methylphenyl)-4-(3-pyridyl)-2-pyrimidineamine, 4-Methyl-N3-[4-(3-pyridin-3-yl)pyrimidin-2-yl]benzene-1,3-diamine, Imatinib Imp F (EP), 4-Methyl-N3-[4-(pyridin-3-yl)pyrimidin-2-yl]benzene-1,3-diamine
IUPAC Name 4-methyl-3-N-(4-pyridin-3-ylpyrimidin-2-yl)benzene-1,3-diamine
SMILES Cc1ccc(N)cc1Nc2nccc(n2)c3cccnc3

Specifications

Quantity 5 g
InChI Formula InChI=1S/C16H15N5/c1-11-4-5-13(17)9-15(11)21-16-19-8-6-14(20-16)12-3-2-7-18-10-12/h2-10H,17H2,1H3,(H,19,20,21)
Formula Weight 277.1327
Chemical Name or Material N-(5-Amino-2-methylphenyl)-4-(3-pyridyl)-2-pyrimidineamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.