missing translation for 'onlineSavingsMsg'
Learn More

N-(5-Chloro-2,1,3-benzothiadiazol-4-yl)-thiourea, TRC

Product Code. 30363954
Change view
Click to view available options
Quantity:
50 mg
500 mg
Unit Size:
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30363954 500 mg 500mg
30368268 50 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30363954 Supplier Toronto Research Chemicals Supplier No. TRCC364995500MG
 Restricted Item
Estimated Shipment: 06-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 51323-05-8
Molecular Formula C7H5ClN4S2
Molecular Weight (g/mol) 244.72
Synonym (5-Chloro-2,1,3-benzothiadiazol-4-yl)-thiourea, Tizanidine EP Impurity B
IUPAC Name (5-chloro-2,1,3-benzothiadiazol-4-yl)thiourea
SMILES NC(=S)Nc1c(Cl)ccc2nsnc12

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C7H5ClN4S2/c8-3-1-2-4-6(12-14-11-4)5(3)10-7(9)13/h1-2H,(H3,9,10,13)
Formula Weight 243.9644
Chemical Name or Material N-(5-Chloro-2,1,3-benzothiadiazol-4-yl)-thiourea
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.