Learn More
N,5-Dimethyl-1-phenyl-(1H-pyrazol-4-ylmethyl)amine, 97%, Thermo Scientific™
Product Code. 10746455
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10746455 250 mg Amber Glass Bottle 250mg
10011364 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10746455 Supplier Thermo Scientific Maybridge Supplier No. CC15546CB
 Restricted Item
Estimated Shipment: 04-11-2026
Add to basket
missing translation for 'validMainframeMsg'
Chemical Identifiers
CAS 1031843-22-7
Molecular Formula C12H15N3
Molecular Weight (g/mol) 201.27
MDL Number MFCD11109316
InChI Key PYMKNYBFTGMBJN-UHFFFAOYSA-N
Synonym methyl 5-methyl-1-phenylpyrazol-4-yl methyl amine, 4-methyl aminomethyl-5-methyl-1-phenyl-1h-pyrazole, n-methyl-1-5-methyl-1-phenyl-1h-pyrazol-4-yl methylamine, methyl 5-methyl-1-phenyl-1h-pyrazol-4-yl methyl amine, n,5-dimethyl-1-phenyl-1h-pyrazol-4-ylmethyl amine, n-methyl-1-5-methyl-1-phenyl-1h-pyrazol-4-yl methanamine
PubChem CID 33589450
IUPAC Name N-methyl-1-(5-methyl-1-phenylpyrazol-4-yl)methanamine
SMILES CNCC1=C(C)N(N=C1)C1=CC=CC=C1
Specifications
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 201.27
Percent Purity 97%
Chemical Name or Material N, 5-Dimethyl-1-phenyl-(1H-pyrazol-4-ylmethyl)amine