missing translation for 'onlineSavingsMsg'
Learn More

N-[6-Chloro-5-(2-methoxyphenoxy)-(2,2-bipyrimidine)4-4-yl]-(1,1-dimethylethyl)-benzenesulfonamide, TRC

Product Code. 30133656
Change view
Click to view available options
Quantity:
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30133656 25 mg 25mg
30009488 50 mg 50mg
30009320 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30133656 Supplier Toronto Research Chemicals Supplier No. TRCC36875025MG
 Restricted Item
Estimated Shipment: 28-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 150727-06-3
Molecular Formula C25 H24 Cl N5 O4 S
Molecular Weight (g/mol) 526.01
Synonym Benzenesulfonamide, N-[6-chloro-5-(2-methoxyphenoxy)[2,2'-bipyrimidin]-4-yl]-4-(1,1-dimethylethyl)-, 4-tert-Butyl-N-[6-chloro-5-(2-methoxyphenoxy)-2,2'-bipyrimidin-4-yl]benzenesulfonamide, 4-tert-Butyl-N-[6-chloro-5-(2-methoxyphenoxy)-2-(2-pyrimidinyl)-4-pyrimidinyl]benzenesulfonamide, 4-tert-Butyl-N-[6-chloro-5-(o-methoxyphenoxy)-2-(2-pyrimidinyl)-4-pyrimidinyl]benzenesulfonamide, N-[6-Chloro-5-(2-methoxyphenoxy)[2,2'-bipyrimidin]-4-yl]-4-(1,1-dimethylethyl)benzenesulfonamide, p-tert-Butyl-N-[6-chloro-5-(o-methoxyphenoxy)-2-(2-pyrimidinyl)-4-pyrimidinyl]benzenesulfonamide, Bosentan Related Compound A
IUPAC Name 4-tert-butyl-N-[6-chloro-5-(2-methoxyphenoxy)-2-pyrimidin-2-ylpyrimidin-4-yl]benzenesulfonamide
SMILES COc1ccccc1Oc2c(Cl)nc(nc2NS(=O)(=O)c3ccc(cc3)C(C)(C)C)c4ncccn4

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C25H24ClN5O4S/c1-25(2,3)16-10-12-17(13-11-16)36(32,33)31-22-20(35-19-9-6-5-8-18(19)34-4)21(26)29-24(30-22)23-27-14-7-15-28-23/h5-15H,1-4H3,(H,29,30,31)
Formula Weight 525.1238
Chemical Name or Material 4-tert-Butyl-N-[6-chloro-5-(2-methoxyphenoxy)-2,2'-bipyrimidin-4-yl]benzenesulfonamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.