missing translation for 'onlineSavingsMsg'
Learn More

N-Acetonylpyridinium chloride, 97%

Product Code. 11468636
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11468636 25 g 25g
11458636 5 g 5g
11478636 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11468636 Supplier Thermo Scientific Alfa Aesar Supplier No. A16015.14
 Restricted Item
Estimated Shipment: 18-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 42508-60-1 | C8H10ClNO | 171.62 g/mol

N-Acetonylpyridinium chloride is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Acetonylpyridinium chloride is used as a pharmaceutical intermediate.

Solubility
Soluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents. Store under dry inert gas. It is hygroscopic in nature. Protect from water and humidity.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 42508-60-1
Molecular Formula C8H10ClNO
Molecular Weight (g/mol) 171.62
MDL Number MFCD00031949
InChI Key JHYYWKUENNZTMD-UHFFFAOYSA-M
Synonym 1-acetonylpyridinium chloride, n-acetonylpyridinium chloride, 1-2-oxopropyl pyridinium chloride, 1-2-oxopropyl pyridin-1-ium chloride, acmc-209jpw, 1-acetonylpyridiniumchloride, 1-pyridin-1-ium-1-ylpropan-2-one chloride, 1-2-oxopropyl-2h-pyridin-2-ylium chloride
PubChem CID 2734121
SMILES [Cl-].CC(=O)C[N+]1=CC=CC=C1

Specifications

Quantity 25 g
Sensitivity Hygroscopic
Solubility Information Soluble in water.
IUPAC Name 1-pyridin-1-ium-1-ylpropan-2-one;chloride
Formula Weight 171.63
Percent Purity 97%
Chemical Name or Material N-Acetonylpyridinium chloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.