missing translation for 'onlineSavingsMsg'
Learn More

N-Acetyl-4-aminosalicylic Acid, TRC

Product Code. 30137532
Change view
Click to view available options
Quantity:
500 mg
1 g
10 g
Unit Size:
10g
1g
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30137532 1 g 1g
30133438 10 g 10g
17944245 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30137532 Supplier Toronto Research Chemicals Supplier No. TRCA1683251G
 Restricted Item
Estimated Shipment: 28-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 50-86-2
Molecular Formula C9 H9 N O4
Molecular Weight (g/mol) 195.17
Synonym 4-(Acetylamino)-2-hydroxybenzoic Acid, Metoclopramide Imp. H (EP)
IUPAC Name 4-acetamido-2-hydroxybenzoic acid
SMILES CC(=O)Nc1ccc(C(=O)O)c(O)c1

Specifications

Quantity 1 g
InChI Formula InChI=1S/C9H9NO4/c1-5(11)10-6-2-3-7(9(13)14)8(12)4-6/h2-4,12H,1H3,(H,10,11)(H,13,14)
Formula Weight 195.0532
Percent Purity >95%
Chemical Name or Material 4-(Acetylamino)-2-hydroxybenzoic Acid
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.