missing translation for 'onlineSavingsMsg'
Learn More

N-Acetyl Adamantamine, TRC

Product Code. 30006784
Change view
Click to view available options
Quantity:
250 mg
1 g
10 g
Unit Size:
10g
1g
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30006784 10 g 10g
17917494 250 mg 250mg
17978714 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30006784 Supplier Toronto Research Chemicals Supplier No. TRCA16805010G
 Restricted Item
Estimated Shipment: 06-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 880-52-4
Molecular Formula C12 H19 N O
Molecular Weight (g/mol) 193.285
Synonym 1-Acetamidoadamantane, Acetamide, N-tricyclo[3.3.1.13,7]dec-1-yl- (9CI, ACI), Acetamide, N-1-adamantyl- (6CI, 7CI, 8CI), N-Tricyclo[3.3.1.13,7]dec-1-ylacetamide (ACI), 1-Acetamidoadamantane, N-(Adamantan-1-yl)acetamide, N-(Tricyclo[3.3.1.13,7]dec-1-yl)ace
IUPAC Name N-(1-adamantyl)acetamide
SMILES CC(=O)NC12CC3CC(CC(C3)C1)C2

Specifications

Quantity 10 g
InChI Formula InChI=1S/C12H19NO/c1-8(14)13-12-5-9-2-10(6-12)4-11(3-9)7-12/h9-11H,2-7H2,1H3,(H,13,14)
Formula Weight 193.147
Chemical Name or Material N-Acetyl Adamantamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.