missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Acetyl-d3-L-4-hydroxyphenylalanine, CDN
Click to view available options
Quantity:
0.005 g
0.01 g
Unit Size:
0.01g
Specifications
Specifications
| Chemical Name or Material | N-Acetyl-L-tyrosine-D3 (N-acetyl-D3) |
| Molecular Formula | C11 D3 H10 N O4 |
| Quantity | 0.01 g |
| Synonym | N-(Acetyl-d3)-L-tyrosine, N-Acetyl-L-tyrosine-D3 (N-acetyl-D3), N-Acetyl-L-tyrosine-d3, (2 S)-2-(2,2,2-Trideuterioacetamido)-3-(4-hydroxyphenyl)propanoic acid, (S)-2-(Acet-d3-amido)-3-(4-hydroxyphenyl)propanoic acid, Ac-d3-L-Tyr-OH, N-(Acetyl-d3)-L-4-hydroxyphenylalanine |
| InChI Formula | InChI=1 S/C11H13NO4/c1-7(13)12-10(11(15)16)6-8-2-4-9(14)5-3-8/h2-5,10,14 H,6H2,1H3,(H,12,13)(H,15,16)/t10-/m0/s1/i1D3 |
| SMILES | [2 H]C([2 H])([2 H])C(=O)N[C@@H](Cc1ccc(O)cc1)C(=O)O |
| IUPAC Name | (2 S)-3-(4-hydroxyphenyl)-2-[(2,2,2-trideuterioacetyl)amino]propanoic acid |
| Molecular Weight (g/mol) | 226.244 |
| Formula Weight | 226.103 g/mol |
| Percent Purity | 98 atom % D, min 98% Chemical Purity |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction