missing translation for 'onlineSavingsMsg'
Learn More

N-Acetyl Dapsone, TRC

Product Code. 17954955
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17954955 10 mg 10mg
17952875 5 mg 5mg
17913885 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17954955 Supplier Toronto Research Chemicals Supplier No. TRCA16843510MG
 Restricted Item
Estimated Shipment: 28-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 565-20-8
Molecular Formula C14 H14 N2 O3 S
Molecular Weight (g/mol) 290.34
Synonym N-Acetyldapsone, Acetanilide, 4'-sulfanilyl- (6CI,7CI,8CI), N-[4-[(4-Aminophenyl)sulfonyl]phenyl]acetamide, 4-Acetamido-4'-aminodiphenyl sulfone, 4-Acetamidophenyl 4'-aminophenyl sulfone, 4-Acetylamino-4'-aminodiphenyl sulfone, 4-Acetylamino-4'-aminophenyl sulfone, 4-Amino-4'-acetamidodiphenyl sulfone, 4'-Sulfanilylacetanilide, Acetyldapsone, MADDS, Monoacetyldapsone, N-Acetyldapsone, NSC 27184, Acetamide, N-[4-[(4-aminophenyl)sulfonyl]phenyl]-
IUPAC Name N-[4-(4-aminophenyl)sulfonylphenyl]acetamide
SMILES CC(=O)Nc1ccc(cc1)S(=O)(=O)c2ccc(N)cc2

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C14H14N2O3S/c1-10(17)16-12-4-8-14(9-5-12)20(18,19)13-6-2-11(15)3-7-13/h2-9H,15H2,1H3,(H,16,17)
Formula Weight 290.0725
Percent Purity >95%
Chemical Name or Material N-Acetyl Dapsone
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.