missing translation for 'onlineSavingsMsg'
Learn More

N-Acetyl Glyphosate, TRC

Product Code. 17924445
Change view
Click to view available options
Quantity:
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17924445 50 mg 50mg
17954595 25 mg 25mg
17940415 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17924445 Supplier Toronto Research Chemicals Supplier No. TRCA17824550MG
 Restricted Item
Estimated Shipment: 20-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 129660-96-4
Molecular Formula C5 H10 N O6 P
Molecular Weight (g/mol) 211.11
Synonym Glycine, N-acetyl-N-(phosphonomethyl)-, N-Acetyl-N-(phosphonomethyl)glycine, N-Acetylglyphosate
IUPAC Name 2-[acetyl(phosphonomethyl)amino]acetic acid
SMILES CC(=O)N(CC(=O)O)CP(=O)(O)O

Specifications

Analyte or Component Names N-Acetyl Glyphosate
Shipping Condition Room Temperature
InChI Formula InChI=1S/C5H10NO6P/c1-4(7)6(2-5(8)9)3-13(10,11)12/h2-3H2,1H3,(H,8,9)(H2,10,11,12)
Solution Type Neat
Industry Type Forensics
Quantity 50 mg
Formula Weight 211.0246
Percent Purity >95%
Chemical Name or Material N-Acetyl Glyphosate
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.