missing translation for 'onlineSavingsMsg'
Learn More

N-Acetyl-L-methionine, 99%

Product Code. 11465737
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 11465737

Brand: Thermo Scientific Alfa Aesar B24312.22

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products

This item is not returnable. View return policy

CAS: 65-82-7 | C7H13NO3S | 191.25 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 65-82-7
Molecular Formula C7H13NO3S
Molecular Weight (g/mol) 191.25
MDL Number MFCD00064441
InChI Key XUYPXLNMDZIRQH-LURJTMIESA-N
Synonym n-acetyl-l-methionine, n-acetylmethionine, acetyl-l-methionine, ac-met-oh, l-n-acetyl methionine, acetylmethionine, methionamine, methionine, n-acetyl-, l, thiomedon, l-methionine, n-acetyl
PubChem CID 448580
ChEBI CHEBI:21557
IUPAC Name (2S)-2-acetamido-4-methylsulfanylbutanoic acid
SMILES CSCC[C@H](NC(C)=O)C(O)=O

Specifications

Melting Point 103°C to 107°C
Beilstein 1725552
Merck Index 14,96
Quantity 100 g
Formula Weight 191.25
Optical Rotation −21° (c=1.3 in Water)
Percent Purity 99%
Chemical Name or Material N-Acetyl-L-methionine

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.