missing translation for 'onlineSavingsMsg'
Learn More

N-Acetyl-S-(2-carboxyethyl)-L-cysteine Bis(dicyclohexylamine) Salt, TRC

Product Code. 30011045
Change view
Click to view available options
Quantity:
10 mg
100 mg
Unit Size:
100mg
10mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30011045 10 mg 10mg
30004428 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30011045 Supplier Toronto Research Chemicals Supplier No. TRCA16804510MG
 Restricted Item
Estimated Shipment: 10-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 201666-27-5
Molecular Formula 2 C12 H23 N . C8 H13 N O5 S
Molecular Weight (g/mol) 597.89
Synonym N-Acetyl-3-(2-carboxyethylthio)alanine Bis(dicyclohexylamine) Salt, 3-(N-Acetyl-L-cystein-S-yl)propanoic Acid Bisdicyclohexylamine) Salt, N-Acetyl-S-(2-carboxyethyl)cysteine Bis(dicyclohexylamine) Salt;
IUPAC Name (2R)-2-acetamido-3-(2-carboxyethylsulfanyl)propanoic acid;N-cyclohexylcyclohexanamine
SMILES CC(=O)N[C@@H](CSCCC(=O)O)C(=O)O.C1CCC(CC1)NC2CCCCC2.C3CCC(CC3)NC4CCCCC4

Specifications

InChI Formula InChI=1S/2C12H23N.C8H13NO5S/c2*1-3-7-11(8-4-1)13-12-9-5-2-6-10-12;1-5(10)9-6(8(13)14)4-15-3-2-7(11)12/h2*11-13H,1-10H2;6H,2-4H2,1H3,(H,9,10)(H,11,12)(H,13,14)/t;;6-/m..0/s1
Quantity 10 mg
Formula Weight 597.4175
Percent Purity >95%
Shipping Condition Room Temperature
Chemical Name or Material N-Acetyl-S-(2-carboxyethyl)-L-cysteine Bis(dicyclohexylamine) Salt
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.