missing translation for 'onlineSavingsMsg'
Learn More

N-Acetyl-S-(2-hydroxypropyl)cysteine Dicyclohexylammonium Salt, TRC

Product Code. 30007964
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30007964 25 mg 25mg
17963585 5 mg 5mg
17983155 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30007964 Supplier Toronto Research Chemicals Supplier No. TRCA17906025MG
 Restricted Item
Estimated Shipment: 11-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 38130-86-8
Molecular Formula C12 H23 N . C8 H15 N O4 S
Molecular Weight (g/mol) 402.59
Synonym L-Cysteine, N-acetyl-S-(2-hydroxypropyl)-, compd. with N-cyclohexylcyclohexanamine (1:1), N-Acetyl-S-(2-hydroxypropyl)-L-cysteine dicyclohexylamine salt, 2-Hydroxypropylmercapturic acid dicyclohexylamine salt, N-Acetyl-S-(2-hydroxypropyl)cysteine dicyclohexylamine, (2R)-2-Acetamido-3-(2-hydroxypropyl)sulfanylpropanoic acid dicyclohexylamine salt
IUPAC Name (2R)-2-acetamido-3-(2-hydroxypropylsulfanyl)propanoic acid;N-cyclohexylcyclohexanamine
SMILES CC(O)CSC[C@H](NC(=O)C)C(=O)O.C1CCC(CC1)NC2CCCCC2

Specifications

InChI Formula InChI=1S/C12H23N.C8H15NO4S/c1-3-7-11(8-4-1)13-12-9-5-2-6-10-12;1-5(10)3-14-4-7(8(12)13)9-6(2)11/h11-13H,1-10H2;5,7,10H,3-4H2,1-2H3,(H,9,11)(H,12,13)/t;5?,7-/m.0/s1
Quantity 25 mg
Formula Weight 402.2552
Percent Purity >95%
Shipping Condition Room Temperature
Chemical Name or Material N-Acetyl-S-(2-hydroxypropyl)cysteine dicyclohexylammonium salt
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.