missing translation for 'onlineSavingsMsg'
Learn More

N-Acetylaminomethylphosphoric Acid (>90%), TRC

Product Code. 30132945
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30132945 5 g 5g
17994065 250 mg 250mg
17913705 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30132945 Supplier Toronto Research Chemicals Supplier No. TRCA1682555G
 Restricted Item
Estimated Shipment: 28-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 57637-97-5
Molecular Formula C3 H8 N O4 P
Molecular Weight (g/mol) 153.07
Synonym Phosphonic acid, (acetamidomethyl)- (6CI), Phosphonic acid, [(acetylamino)methyl]- (9CI), P-[(Acetylamino)methyl]phosphonic acid, N-Acetylaminomethylphosphonic acid, Acetamidomethylphosphonic Acid
IUPAC Name acetamidomethylphosphonic acid
SMILES CC(=O)NCP(=O)(O)O

Specifications

Quantity 5 g
InChI Formula InChI=1S/C3H8NO4P/c1-3(5)4-2-9(6,7)8/h2H2,1H3,(H,4,5)(H2,6,7,8)
Formula Weight 153.0191
Percent Purity >95%
Chemical Name or Material N-Acetylaminomethylphosphoric Acid
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.