missing translation for 'onlineSavingsMsg'
Learn More

N-Acetylethanolamine (~90%), TRC

Product Code. 30319572
Change view
Click to view available options
Quantity:
500 mg
1 g
5 g
Unit Size:
1g
500mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30319572 5 g 5g
30323654 1 g 1g
30322040 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30319572 Supplier Toronto Research Chemicals Supplier No. TRCA2848305G
 Restricted Item
Estimated Shipment: 06-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N-Acetylethanolamine
CAS 142-26-7
Molecular Formula C4 H9 N O2
Quantity 5 g
Synonym N-(2-Hydroxyethyl)acetamide, 2-Acetamidoethanol, 2-Acetylaminoethanol, 2-N-Acetylaminoethanol, Acetamide MEA, Acetylcolamine, Incromectant AMEA 100, Incromectant AMEA 70, Mackamide AME 100, N-(β-Hydroxyethyl)acetamide, N-Acetyl-2-aminoethanol, N-Ethanolacetamide, NSC 5999, Schercomid AME 70, β-Hydroxyethylacetamide
InChI Formula InChI=1S/C4H9NO2/c1-4(7)5-2-3-6/h6H,2-3H2,1H3,(H,5,7)
SMILES CC(=O)NCCO
IUPAC Name N-(2-hydroxyethyl)acetamide
Molecular Weight (g/mol) 103.12
Formula Weight 103.0633
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.