missing translation for 'onlineSavingsMsg'
Learn More

N-Acetylsulfamerazine, TRC

Product Code. 30133273
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30133273 50 mg 50mg
17980355 10 mg 10mg
17923965 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30133273 Supplier Toronto Research Chemicals Supplier No. TRCA18782050MG
 Restricted Item
Estimated Shipment: 06-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 127-73-1
Molecular Formula C13H14N4O3S
Molecular Weight (g/mol) 306.34
Synonym Sulfamerazine-N-acetyl, Acetanilide, 4'-[(4-methyl-2-pyrimidinyl)sulfamoyl]- (6CI,7CI,8CI), N-[4-[[(4-Methyl-2-pyrimidinyl)amino]sulfonyl]phenyl]acetamide, 4'-[(4-Methyl-2-pyrimidinyl)sulfamoyl]acetanilide, Acetylsulfamerazine, N4-Acetylsulfamerazine, Sulfamelazine
IUPAC Name N-[4-[(4-methylpyrimidin-2-yl)sulfamoyl]phenyl]acetamide
SMILES CC(=O)Nc1ccc(cc1)S(=O)(=O)Nc2nccc(C)n2

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C13H14N4O3S/c1-9-7-8-14-13(15-9)17-21(19,20)12-5-3-11(4-6-12)16-10(2)18/h3-8H,1-2H3,(H,16,18)(H,14,15,17)
Formula Weight 306.0787
Chemical Name or Material N-Acetylsulfamerazine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.