missing translation for 'onlineSavingsMsg'
Learn More

N-Acetyltizanidine, Mikromol™

Product Code. 17992436
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17992436 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17992436 Supplier Mikromol Supplier No. MM0887.04
 Restricted Item
Estimated Shipment: 02-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

N-Acetyltizanidine
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N-Acetyltizanidine
CAS 173532-15-5
Analyte or Component Names N-Acetyltizanidine
Shipping Condition Room Temperature
Molecular Formula C11 H10 Cl N5 O S
Certifications/Compliance ISO 17025
Synonym N-Acetyltizanidine, Tizanidine Related Compound B
InChI Formula InChI=1S/C11H10ClN5OS/c1-6(18)17-5-4-13-11(17)14-9-7(12)2-3-8-10(9)16-19-15-8/h2-3H,4-5H2,1H3,(H,13,14)
SMILES CC(=O)N1CCN=C1Nc2c(Cl)ccc3nsnc23
IUPAC Name 1-[2-[(5-chloro-2,1,3-benzothiadiazol-4-yl)amino]-4,5-dihydroimidazol-1-yl]ethanone
Molecular Weight (g/mol) 295.75
Quantity 50 mg
Formula Weight 295.0295 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.