Learn More
N-Alkyl(C12-16)-N-Benzyl-N,N-Dimethylammonium Chloride (Technical Grade), TRC
Product Code. 30011424
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
10g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30011424 5 g 5g
30008092 10 g 10g
30006432 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Regulatory: This product fall under regulation “Lov om kemiske stoffer og produkter - lovbekendtgørelse nr. 1755 af 22. december 2006. If applicable, customers must provide a license before delivery.
Product Code. 30011424 Supplier Toronto Research Chemicals Supplier No. TRCA5751385G
 Restricted Item
This item requires a price update prior to being ordered.
Request a Price for this item here.
missing translation for 'validMainframeMsg'
Regulatory: This product fall under regulation “Lov om kemiske stoffer og produkter - lovbekendtgørelse nr. 1755 af 22. december 2006. If applicable, customers must provide a license before delivery.
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 68424-85-1
Molecular Formula C17H30ClN
Molecular Weight (g/mol) 283.88
IUPAC Name benzyl-dimethyl-tetradecylazanium;chloride
SMILES CCCCCCCCCCCCCC[N+](C)(C)CC1=CC=CC=C1.[Cl-]
Specifications
Quantity 5 g
InChI Formula InChI=1S/C23H42N.ClH/c1-4-5-6-7-8-9-10-11-12-13-14-18-21-24(2,3)22-23-19-16-15-17-20-23;/h15-17,19-20H,4-14,18,21-22H2,1-3H3;1H/q+1;/p-1
Chemical Name or Material N-Alkyl(C12-16)-N-Benzyl-N,N-Dimethylammonium Chloride