missing translation for 'onlineSavingsMsg'
Learn More

N-alpha-(2,4-Dinitro-5-fluorophenyl)-L-alaninamide, 98%

Product Code. 11938602 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 mg
Packaging:
Glass bottle
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
11938602 100 mg Glass bottle 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11938602 Supplier Thermo Scientific Acros Supplier No. 439341000
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternativa produkter

missing translation for 'validMainframeMsg'

Denna artikel kan inte returneras. Se returpolicy

CAS: 95713-52-3 | C9H9FN4O5 | 272.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chiral derivatization reagent
TRUSTED_SUSTAINABILITY

Specifikationer

CAS 95713-52-3
CAS Min % 97.5
CAS Max % 100.0
Melting Point 227.7°C to 229.4°C
Color Yellow to Orange
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Molecular Formula C9H9FN4O5
MDL Number MFCD00042049
Quantity 100 mg
Specific Rotation +54.0° to +58.0° (20°C, 589nm) (c=1, acetone)
Synonym marfey's reagent, s-2-5-fluoro-2,4-dinitrophenyl amino propanamide, 2s-2-5-fluoro-2,4-dinitrophenyl amino propanamide, n-2,4-dinitro-5-fluorophenyl-l-alaninamide, 2s-2-5-fluoro-2,4-dinitroanilino propanamide, propanamide, 2-5-fluoro-2,4-dinitrophenyl amino-, 2s, nalpha-2,4-dinitro-5-fluorophenyl-l-alaninamide, n-alpha-2,4-dinitro-5-fluorophenyl-l-alaninamide, fdaa, 1-fd-aa
InChI Key NEPLBHLFDJOJGP-BYPYZUCNSA-N
SMILES C[C@H](NC1=CC(F)=C(C=C1[N+]([O-])=O)[N+]([O-])=O)C(N)=O
IUPAC Name (2S)-2-(5-fluoro-2,4-dinitroanilino)propanamide
Molecular Weight (g/mol) 272.19
PubChem CID 5486955
Formula Weight 272.19
Percent Purity 98%
Physical Form Powder
Chemical Name or Material N-α-(2, 4-Dinitro-5-fluorophenyl)-L-alaninamide
Visa mer Visa mindre
Produktbeteckning
  • N-alpha-(2,4-Dinitro-5-fluorophenyl)-L-alaninamide, 98%

RUO – Research Use Only

Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.