missing translation for 'onlineSavingsMsg'
Learn More

N-Benzyl-3-hydroxypiperidine, TRC

Product Code. 30330268
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Quantity:
1 g
missing translation for 'unitSize':
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity missing translation for 'unitSize'
30330268 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 missing translation for 'options'
This item is not returnable. View return policy
Product Code. 30330268 missing translation for 'mfr' Toronto Research Chemicals missing translation for 'supplierNo' TRCB2788201G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N-Benzyl-3-hydroxypiperidine
CAS 14813-01-5
Molecular Formula C12 H17 N O
Quantity 1 g
Synonym 3-Piperidinol, 1-(phenylmethyl)-, 3-Piperidinol, 1-benzyl- (7CI,8CI), 1-(Phenylmethyl)-3-piperidinol, 1-Benzyl-3-hydroxypiperidine, 1-Benzyl-3-piperidinol, 3-Hydroxy-N-benzylpiperidine, N-Benzyl-3-hydroxypiperidine, N-Benzyl-3-piperidinol, NSC 111182
InChI Formula InChI=1S/C12H17NO/c14-12-7-4-8-13(10-12)9-11-5-2-1-3-6-11/h1-3,5-6,12,14H,4,7-10H2
SMILES OC1CCCN(Cc2ccccc2)C1
IUPAC Name 1-benzylpiperidin-3-ol
Molecular Weight (g/mol) 191.27
Formula Weight 191.131
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.