missing translation for 'onlineSavingsMsg'
Learn More

N-Benzyl Salbutamol Acetonide, TRC

Product Code. 30330943
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30330943 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30330943 Supplier Toronto Research Chemicals Supplier No. TRCB289020250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N-Benzyl Salbutamol Acetonide
CAS 1797114-57-8
Molecular Formula C23H31NO3
Quantity 250 mg
Synonym α-[[(1,1-Dimethylethyl)(phenylmethyl)amino]methyl]-2,2-dimethyl-4H-1,3-benzodioxin-6-methanol, N-Benzyl Salbutamol Acetonide
InChI Formula InChI=1S/C23H31NO3/c1-22(2,3)24(14-17-9-7-6-8-10-17)15-20(25)18-11-12-21-19(13-18)16-26-23(4,5)27-21/h6-13,20,25H,14-16H2,1-5H3
SMILES CC(C)(C)N(CC(O)c1ccc2OC(C)(C)OCc2c1)Cc3ccccc3
IUPAC Name 2-[benzyl(tert-butyl)amino]-1-(2,2-dimethyl-4H-1,3-benzodioxin-6-yl)ethanol
Molecular Weight (g/mol) 369.4971
Formula Weight 369.2304
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.