missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-4-oxo-L-proline, 98%, Thermo Scientific™

Product Code. 11446190
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11446190 5 g 5g
15483607 250 mg 250mg
11436190 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11446190 Supplier Thermo Scientific Alfa Aesar Supplier No. H27104.06

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
N-Boc-4-oxo-L-proline is used in practical Synthesis of Boc-Protected cis-4-Trifluoromethyl and cis-4-Difluoromethyl-l- prolines.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 84348-37-8
Molecular Formula C10H15NO5
Molecular Weight (g/mol) 229.23
MDL Number MFCD01860669
InChI Key CKYGSXRXTIKGAJ-UHFFFAOYNA-N
Synonym n-boc-4-oxo-l-proline, s-1-tert-butoxycarbonyl-4-oxopyrrolidine-2-carboxylic acid, n-t-boc-4-oxo-l-proline, boc-4-oxo-pro-oh, n-tert-boc-4-oxo-l-proline, 2s-1-tert-butoxycarbonyl-4-oxopyrrolidine-2-carboxylic acid, 1-tert-butoxycarbonyl-4-oxoproline, 2s-1-tert-butoxycarbonyl-4-oxo-pyrrolidine-2-carboxylic acid, 2s-1-tert-butoxy carbonyl-4-oxopyrrolidine-2-carboxylic acid, s-4-oxo-pyrrolidine-1,2-dicarboxylic acid 1-tert-butyl ester
PubChem CID 11593804
IUPAC Name (2S)-1-[(2-methylpropan-2-yl)oxycarbonyl]-4-oxopyrrolidine-2-carboxylic acid
SMILES CC(C)(C)OC(=O)N1CC(=O)CC1C(O)=O

Specifications

Melting Point ∽160°C (decomposition)
Quantity 5 g
Solubility Information Insoluble in water.
Formula Weight 229.24
Percent Purity 98%
Chemical Name or Material N-Boc-4-oxo-L-proline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.