missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-glutamic Acid Tert-butyl Ester, TRC

Product Code. 30329992
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30329992 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30329992 Supplier Toronto Research Chemicals Supplier No. TRCB6561355G
 Restricted Item
Estimated Shipment: 06-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N-Boc-glutamic Acid Tert-butyl Ester
CAS 24277-39-2
Molecular Formula C14H25NO6
Quantity 5 g
Synonym (4S)-5-tert-Butoxy-4-[(tert-butoxycarbonyl)amino]-5-oxopentanoic Acid, (S)-2-[(tert-Butoxycarbonyl)amino]pentanedioic Acid 1-Tert-butyl Ester, (S)-5-tert-Butoxy-4-(tert-butoxycarbonylamino)-5-oxopentanoic Acid, BOC-L-glutamic Acid Tert-butyl Ester, N-(tert-Butoxycarbonyl)-L-glutamic Acid 1-Tert-butyl Ester, N-tert-Butoxycarbonylglutamic Acid alpha-Tert-butyl Ester, N-alpha-tert-Butoxycarbonyl-L-glutamic Acid alpha-Tert Butyl Ester, NSC 164659
InChI Formula InChI=1S/C14H25NO6/c1-13(2,3)20-11(18)9(7-8-10(16)17)15-12(19)21-14(4,5)6/h9H,7-8H2,1-6H3,(H,15,19)(H,16,17)/t9-/m0/s1
SMILES CC(C)(C)OC(=O)N[C@@H](CCC(=O)O)C(=O)OC(C)(C)C
IUPAC Name (4S)-5-[(2-methylpropan-2-yl)oxy]-4-[(2-methylpropan-2-yl)oxycarbonylamino]-5-oxopentanoic acid
Molecular Weight (g/mol) 303.35
Formula Weight 303.1682
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.