missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-hydroxylamine, TRC

Product Code. 30337259
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30337259 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30337259 Supplier Toronto Research Chemicals Supplier No. TRCB65647025G
 Restricted Item
Estimated Shipment: 31-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N-Boc-hydroxylamine
CAS 36016-38-3
Molecular Formula C5H11NO3
Quantity 25 g
Synonym N-Hydroxycarbamic Acid 1,1-Dimethylethyl Ester, Hydroxycarbamic Acid 1,1-Dimethylethyl Ester, Hydroxycarbamic Acid tert-Butyl Ester, 1,1-Dimethylethyl N-Hydroxycarbamate, N-(tert-Butoxycarbonyl)hydroxylamine, N-(tert-Butyloxycarbonyl)hydroxylamine, N-Boc-hydroxylamine, N-Hydroxycarbamic Acid tert-Butyl Ester, N-t-Butyloxycarbonyl Hydroxylamine, NSC 131086, NSC 144620, tert-Butoxycarbonylhydroxylamine, tert-Butoxyhydroxamic Acid, tert-Butyl N-Hydroxycarbamate, tert-Butyl Hydroxycarbamate
InChI Formula InChI=1S/C5H11NO3/c1-5(2,3)9-4(7)6-8/h8H,1-3H3,(H,6,7)
SMILES CC(C)(C)OC(=O)NO
IUPAC Name tert-butyl N-hydroxycarbamate
Molecular Weight (g/mol) 133.15
Formula Weight 133.0739
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.