missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-L-alanine, 98+%

Product Code. p-7023000
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 11408646

Brand: Thermo Scientific Alfa Aesar A16018.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 15761-38-3 | C8H15NO4 | 189.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

 
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15761-38-3
Molecular Formula C8H15NO4
Molecular Weight (g/mol) 189.21
MDL Number MFCD00037225
InChI Key QVHJQCGUWFKTSE-YFKPBYRVSA-N
Synonym boc-ala-oh, n-tert-butoxycarbonyl-l-alanine, boc-l-alanine, n-boc-l-alanine, boc-l-ala-oh, n-t-boc-l-alanine, boc-ala, tert-butoxycarbonyl-l-alanine, tert-butoxycarbonyl alanine, n-alpha-t-boc-l-alanine
PubChem CID 85082
IUPAC Name (2S)-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid
SMILES C[C@H](NC(=O)OC(C)(C)C)C(O)=O

Specifications

Melting Point 80°C to 84°C
Quantity 5 g
Beilstein 1726365
Optical Rotation −26° (c=2 in Acetic Acid)
Formula Weight 189.21
Percent Purity ≥98%
Chemical Name or Material N-Boc-L-alanine

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.