missing translation for 'onlineSavingsMsg'
Learn More
Learn More
n-Butyl-d9-amine, CDN
Click to view available options
Quantity:
0.25 g
0.5 g
Unit Size:
0.25g
0.5g
Specifications
Specifications
| Chemical Name or Material | 1-Butanamine-d9 |
| CAS | 776285-22-4 |
| Molecular Formula | C4H2D9N |
| Quantity | 0.25 g |
| Synonym | n-Butyl-d9-amine, 1-Aminobutane-d9, 1-Butylamine-d9, Mono-n-butylamine-d9, Monobutylamine-d9, NSC 8029-d9, Norvalamine-d9, n-Butylamine-d9; |
| InChI Formula | InChI=1 S/C4H11N/c1-2-3-4-5/h2-5H2,1H3/i1D3,2D2,3D2,4D2 |
| SMILES | NC([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])[2 H] |
| IUPAC Name | butan-d9-1-amine |
| Molecular Weight (g/mol) | 82.19 |
| Formula Weight | 82.15 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction