Learn More
n-Butyl formate, 97%
Product Code. 11438760
Change view
Click to view available options
Quantity:
50 mL
250 mL
1000 mL
Unit Size:
1000mL
250mL
50mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11438760 50 mL 50mL
11431578 250 mL 250mL
11421578 1000 mL 1000mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11438760 Supplier Thermo Scientific Alfa Aesar Supplier No. A13882.AD
 Restricted Item
In Stock
Add to basket
missing translation for 'validMainframeMsg'
CAS: 592-84-7 | C5H10O2 | 102.13 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 592-84-7
Molecular Formula C5H10O2
Molecular Weight (g/mol) 102.13
MDL Number MFCD00003296
InChI Key NMJJFJNHVMGPGM-UHFFFAOYSA-N
Synonym n-butyl formate, formic acid, butyl ester, butyl methanoate, n-butyl methanoate, formic acid butyl ester, n-butylformate, butylester kyseliny mravenci, fema no. 2196, unii-0y9mz7vm9p, butylester kyseliny mravenci czech
PubChem CID 11614
IUPAC Name butyl formate
SMILES CCCCOC=O
Specifications
Melting Point -92°C
Boiling Point 105°C to 107°C
Flash Point 18°C (64°F)
Quantity 50 mL
UN Number UN1128
Beilstein 1742108
Formula Weight 102.13
Percent Purity 97%
Chemical Name or Material n-Butyl formate