missing translation for 'onlineSavingsMsg'
Learn More

n-Butyl levulinate, 98%

Product Code. 15435538
Change view
Click to view available options
Quantity:
100 mL
500 mL
Unit Size:
100mL
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15435538 500 mL 500mL
15425538 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15435538 Supplier Thermo Scientific Alfa Aesar Supplier No. H59274.AP
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 2052-15-5 | C9H16O3 | 172.224 g/mol

Butyl levulinate was used in the synthesis of γ-valerolactone.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Butyl levulinate was used in the synthesis of γ-valerolactone.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2052-15-5
Molecular Formula C9H16O3
Molecular Weight (g/mol) 172.224
MDL Number MFCD00009449
InChI Key ISBWNEKJSSLXOD-UHFFFAOYSA-N
Synonym butyl levulinate, n-butyl levulinate, butyl laevulinate, pentanoic acid, 4-oxo-, butyl ester, n-butyl laevulinate, levulinic acid, butyl ester, butyl 4-ketovalerate, 4-ketopentanoic acid butyl ester, butyl acetylpropionate, n-butyl 4-oxopentanoate
PubChem CID 16331
IUPAC Name butyl 4-oxopentanoate
SMILES CCCCOC(=O)CCC(=O)C

Specifications

Density 0.974
Boiling Point 106°C to 108°C (5.5 mmHg)
Flash Point 92°C (198°F)
Linear Formula CH3COCH2CH2CO2(CH2)3CH3
Refractive Index 1.427
Quantity 500 mL
Solubility Information Slightly soluble in water.
Formula Weight 172.22
Percent Purity 98%
Chemical Name or Material n-Butyl levulinate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.