missing translation for 'onlineSavingsMsg'
Learn More

n-Butyl methyl sulfide, 98+%, Thermo Scientific™

Product Code. p-7045804
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11398575 5 g 5g
11308585 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11398575 Supplier Thermo Scientific Alfa Aesar Supplier No. L12039.06

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 628-29-5
Molecular Formula C5H12S
Molecular Weight (g/mol) 104.21
MDL Number MFCD00015256
InChI Key WCXXISMIJBRDQK-UHFFFAOYSA-N
Synonym butyl methyl sulfide, butane, 1-methylthio, n-butyl methyl sulfide, butyl methyl sulphide, sulfide, butyl methyl, 1-methylthio butane, unii-2n5imb9xap, methyl butyl sulfide, 2-thiahexane, 2n5imb9xap
PubChem CID 12339
IUPAC Name 1-methylsulfanylbutane
SMILES CCCCSC

Specifications

Density 0.85
Boiling Point 122°C to 123°C
Flash Point 20°C (68°F)
Odor Unpleasant
Linear Formula CH3S(CH2)3CH3
Refractive Index 1.4621
Quantity 5 g
UN Number UN1993
Beilstein 1730959
Formula Weight 104.22
Percent Purity ≥98%
Chemical Name or Material n-Butyl methyl sulfide
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.