missing translation for 'onlineSavingsMsg'
Learn More

N-Butylscopolammonium Bromide, TRC

Product Code. 17964615
Change view
Click to view available options
Quantity:
50 mg
1 g
5 g
Unit Size:
1g
50mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17964615 1 g 1g
17922905 5 g 5g
17902035 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17964615 Supplier Toronto Research Chemicals Supplier No. TRCB6937501G
 Restricted Item
Estimated Shipment: 28-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 149-64-4
Molecular Formula C21 H30 N O4 . Br
Molecular Weight (g/mol) 440.37
Synonym Hyoscine Butylbromide, (1α,2β,4β,5α,7β)-9-Butyl-7-[(2S)-3-hydroxy-1-oxo-2-phenylpropoxy]-9-methyl-3-Oxa-9-azoniatricyclo[3.3.1.02,4]nonane bromide (1:1), 8-Butyl-6β,7β-epoxy-3α-hydroxy-1αH,5αH-tropanium bromide (-)-tropate, [7(S)-(1α,2β,4β,5α,7β)]-9-Butyl-7-(3-hydroxy-1-oxo-2-phenylpropoxy)-9-methyl-3-oxa-9-azoniatricyclo[3.3.1.02,4]nonane bromide, N-Butylscopolammonium bromide, (-)-Tropic acid ester with 8-butyl-6β,7β-epoxy-3α-hydroxy-1αH,5αH-tropanium bromide, (-)-Scopolamine butylbromide, Amisepan, Buscapine, Buscol, Buscolamin, Buscolysin, Buscolysine, Buscopan, Butylmin, Butylscopolamine bromide, Butylscopolammonium bromide, Donopon, Hyoscine butobromide, Hyoscine butyl bromide, Joscine, Monospan, N-Butylhyoscine bromide, N-Butylhyoscinium bromide, N-Butylscopolamine bromide, N-Butylscopolaminium bromide, Scobro, Scobron, Scobutil, Scobutyl, Scopolamine bromobutylate, Scopolamine butobromide, Scopolamine butyl bromide, Scopolamine-N-butyl bromide, Scopolan, Sparicon, Sporamin, Sporamin (pharmaceutical), Stibron, Tirantil
IUPAC Name (1α,2β,4β,5α,7β)-9-Butyl-7-[(2S)-3-hydroxy-1-oxo-2-phenylpropoxy]-9-methyl-3-oxa-9-azoniatricyclo[3.3.1.02,4]nonane bromide
SMILES [Br-].CCCC[N@@+]1(C)[C@@H]2C[C@H](C[C@H]1[C@@H]3O[C@H]23)OC(=O)[C@H](CO)c4ccccc4

Specifications

Quantity 1 g
InChI Formula InChI=1S/C21H30NO4.BrH/c1-3-4-10-22(2)17-11-15(12-18(22)20-19(17)26-20)25-21(24)16(13-23)14-8-6-5-7-9-14;/h5-9,15-20,23H,3-4,10-13H2,1-2H3;1H/q+1;/p-1/t15-,16-,17-,18+,19-,20+,22+;/m1./s1
Formula Weight 439.1358
Percent Purity >95%
Chemical Name or Material Hyoscine Butylbromide
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.