missing translation for 'onlineSavingsMsg'
Learn More

n-(Butylthio)acetonitrile, 95%, Thermo Scientific™

Product Code. 11466046
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11466046 5 g 5g
11476046 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11466046 Supplier Thermo Scientific Alfa Aesar Supplier No. B22336.06

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 71037-08-6
Molecular Formula C6H11NS
Molecular Weight (g/mol) 129.22
MDL Number MFCD00041458
InChI Key LRMUBLYDYGZCDR-UHFFFAOYSA-N
Synonym n-butylthio acetonitrile, 2-butylthio acetonitrile, 2-butylsulfanyl acetonitrile, acetonitrile,2-butylthio, butylthio acetonitrile
PubChem CID 7567852
IUPAC Name 2-butylsulfanylacetonitrile
SMILES CCCCSCC#N

Specifications

Boiling Point 134°C to 135°C (40 mmHg)
Linear Formula CH3(CH2)3SCH2CN
Refractive Index 1.472
Quantity 5 g
UN Number UN3276
Beilstein 4174357
Formula Weight 129.23
Percent Purity 95%
Chemical Name or Material n-(Butylthio)acetonitrile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.