missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Cadherin Antibody (CL3716) [Alexa Fluor™ 350], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-61632AF350
This item is not returnable.
View return policy
Description
N-Cadherin Monoclonal antibody specifically detects N-Cadherin in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),KnockDown,Immunocytochemistry/ Immunofluorescence
Specifications
| N-Cadherin | |
| Monoclonal | |
| Alexa Fluor 350 | |
| ACOGS, ARVD14, cadherin 2, type 1, N-cadherin (neuronal), cadherin-2, calcium-dependent adhesion protein, neuronal, CD325, CD325 antigen, CDH2, CDHN, CDw325, NCAD, N-cadherin, N-cadherin 1, Neural cadherin, neural-cadherin | |
| This antibody was developed using a recombinant protein derived from P19022, with the exact immunogen sequence remaining proprietary. | |
| 0.1 mL | |
| Cancer, Cell Cycle and Replication, Cellular Markers, Extracellular Matrix, Mesenchymal Stem Cell Markers, Neuroscience, Plasma Membrane Markers, Signal Transduction, Stem Cells | |
| 1000 | |
| Store at 4⪚C in the dark. | |
| IgG1 |
| Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), KnockDown, Immunocytochemistry/Immunofluorescence | |
| CL3716 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction