missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Cadherin Antibody (GC-4), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-21805
This item is not returnable.
View return policy
Description
N-Cadherin Monoclonal specifically detects N-Cadherin in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen, In vitro assay, Electron Microscopy.
Specifications
| N-Cadherin | |
| Monoclonal | |
| Unconjugated | |
| cadherin 2, N-cadherin (neuronal), cadherin 2, type 1, N-cadherin (neuronal), cadherin-2, CD325, CD325 antigen, CDHNcalcium-dependent adhesion protein, neuronal, CDw325, N-cadherin, NCADN-cadherin 1, Neural cadherin, neural-cadherin | |
| Mouse | |
| 135 kDa | |
| 0.2 mL | |
| Cancer, Cell Cycle and Replication, Cellular Markers, Extracellular Matrix, Mesenchymal Stem Cell Markers, Neuroscience, Plasma Membrane Markers, Signal Transduction, Stem Cells | |
| 1000 | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen), In vitro Assay, Immunoassay | |
| GC-4 | |
| Western Blot 10-20ug/mL, Flow Cytometry 1:50-1:250, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:50-1:250, Immunohistochemistry-Frozen 10-20ug/mL, In vitro assay, Electron Microscopy | |
| CDH2 | |
| This N-Cadherin Antibody (GC-4) was developed against affinity purified chicken heart A-CAM. | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Chicken, Monkey, Rabbit, Human, Mouse, Rat. | |
| Store at -20C. Avoid freeze-thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction