missing translation for 'onlineSavingsMsg'
Learn More

N-Cadherin Antibody (rCDH2/8291), Alexa Fluor™ 350, Novus Biologicals™

Mouse Monoclonal Antibody

Brand:  Novus Biologicals NBP3-23991AF350

Product Code. 18557546

  • 679.00€ / 0.10mL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

N-Cadherin Monoclonal antibody specifically detects N-Cadherin in Human samples. It is validated for Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications

Specifications

N-Cadherin
Monoclonal
Alexa Fluor 350
ACOGS, ARVD14, cadherin 2, type 1, N-cadherin (neuronal), cadherin-2, calcium-dependent adhesion protein, neuronal, CD325, CD325 antigen, CDH2, CDHN, CDw325, NCAD, N-cadherin, N-cadherin 1, Neural cadherin, neural-cadherin
Recombinant fragment (around aa605-905) of human N-Cadherin protein (exact sequence is proprietary)
0.1 mL
Cancer, Cell Cycle and Replication, Cellular Markers, Extracellular Matrix, Mesenchymal Stem Cell Markers, Neuroscience, Plasma Membrane Markers, Signal Transduction, Stem Cells
1000
Store at 4°C in the dark.
IgG1 κ
Immunohistochemistry (Paraffin)
rCDH2/8291
50 mM Sodium Borate
Mouse
Protein A or G purified
RUO
Primary
Human
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.