missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Carbobenzyloxy-L-glutamic Acid O-Benzyl Ester, TRC
Click to view available options
Quantity:
10 g
Unit Size:
10g
Specifications
Specifications
| Chemical Name or Material | N-Carbobenzyloxy-L-glutamic Acid O-Benzyl Ester |
| CAS | 3705-42-8 |
| Molecular Formula | C20H21NO6 |
| Quantity | 10 g |
| Synonym | N-[(Phenylmethoxy)carbonyl]-L-glutamic Acid 1-(Phenylmethyl)ester, N-(Benzyloxycarbonyl)-L-glutamic Acid alpha-Benzyl Ester, N-Benzyloxycarbonyl-L-glutamic Acid 1-Benzyl Ester, NSC 169160, alpha-Benzyl N-(benzyloxycarbonyl)-L-glutamate |
| InChI Formula | InChI=1S/C20H21NO6/c22-18(23)12-11-17(19(24)26-13-15-7-3-1-4-8-15)21-20(25)27-14-16-9-5-2-6-10-16/h1-10,17H,11-14H2,(H,21,25)(H,22,23)/t17-/m0/s1 |
| SMILES | OC(=O)CC[C@H](NC(=O)OCc1ccccc1)C(=O)OCc2ccccc2 |
| IUPAC Name | (4S)-5-oxo-5-phenylmethoxy-4-(phenylmethoxycarbonylamino)pentanoic acid |
| Molecular Weight (g/mol) | 371.38 |
| Formula Weight | 371.1369 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction