missing translation for 'onlineSavingsMsg'
Learn More

N-De(4-sulfonamidophenyl)-N'-(4-sulfonamidophenyl) Celecoxib, TRC

Product Code. 30012619
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30012619 10 mg 10mg
30004831 100 mg 100mg
30012216 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30012619 Supplier Toronto Research Chemicals Supplier No. TRCD29791510MG
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 331943-04-5
Molecular Formula C17 H14 F3 N3 O2 S
Molecular Weight (g/mol) 381.37
Synonym Benzenesulfonamide, 4-[3-(4-methylphenyl)-5-(trifluoromethyl)-1H-pyrazol-1-yl]-, Celecoxib Imp. B (EP), Celecoxib RC B (USP)
IUPAC Name 4-[3-(4-methylphenyl)-5-(trifluoromethyl)pyrazol-1-yl]benzenesulfonamide
SMILES Cc1ccc(cc1)c2cc(n(n2)c3ccc(cc3)S(=O)(=O)N)C(F)(F)F

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C17H14F3N3O2S/c1-11-2-4-12(5-3-11)15-10-16(17(18,19)20)23(22-15)13-6-8-14(9-7-13)26(21,24)25/h2-10H,1H3,(H2,21,24,25)
Formula Weight 381.0759
Percent Purity >95%
Chemical Name or Material 4-[3-(4-methylphenyl)-5-(trifluoromethyl)-1H-pyrazol-1-yl]benzenesulfonamide
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.