missing translation for 'onlineSavingsMsg'
Learn More

N-Dechloroethyl Cyclophosphamide, TRC

Product Code. 30133437
Change view
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30133437 10 mg 10mg
30006989 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30133437 Supplier Toronto Research Chemicals Supplier No. TRCD22640010MG
 Restricted Item
Estimated Shipment: 28-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 36761-83-8
Molecular Formula C5 H12 Cl N2 O2 P
Molecular Weight (g/mol) 198.59
Synonym 2H-1,3,2-Oxazaphosphorin-2-amine, N-(2-chloroethyl)tetrahydro-, 2-oxide, 2-(2-Chloroethylamino)tetrahydro-2H-1,3,2-oxazaphosphorine 2-oxide, 3-Dechloroethylifosfamide, Asta 4968, Dechloroethylcyclophosphamide, Monochloroethylcyclophosphamide, N-Dechloroethylcyclophosphamide, (2RS)-2-[(2-Chloroethyl)amino]-1,3,2λ5-oxazaphosphinan-2-one
IUPAC Name N-(2-chloroethyl)-2-oxo-1,3,2λ^{5}-oxazaphosphinan-2-amine
SMILES ClCCNP1(=O)NCCCO1

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C5H12ClN2O2P/c6-2-4-8-11(9)7-3-1-5-10-11/h1-5H2,(H2,7,8,9)
Formula Weight 198.0325
Chemical Name or Material N-Dechloroethyl Cyclophosphamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.