missing translation for 'onlineSavingsMsg'
Learn More

n-Decyltrichlorosilane, 97%

Product Code. 11397635
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11397635 50 g 50g
11387635 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11397635 Supplier Thermo Scientific Alfa Aesar Supplier No. L05912.18
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 13829-21-5 | C10H21Cl3Si | 275.713 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13829-21-5
Molecular Formula C10H21Cl3Si
Molecular Weight (g/mol) 275.713
MDL Number MFCD00013600
InChI Key HLWCOIUDOLYBGD-UHFFFAOYSA-N
Synonym decyltrichlorosilane, trichloro decyl silane, n-decyltrichlorosilane, silane, trichlorodecyl, acmc-209chi, trichloro n-decyl silane, trichloro decyl silane #
PubChem CID 83764
IUPAC Name trichloro(decyl)silane
SMILES CCCCCCCCCC[Si](Cl)(Cl)Cl

Specifications

Boiling Point 133°C to 137°C (5mmHg)
Quantity 50 g
Linear Formula CH3(CH2)9SiCl3
UN Number UN2987
Beilstein 1749722
Flash Point >110°C (230°F)
Formula Weight 275.72
Percent Purity 97%
Refractive Index 1.452
Sensitivity Moisture sensitive
Density 1.049
Chemical Name or Material n-Decyltrichlorosilane
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.